Learn More
Corning™ Costar™ Cell Culture Triangular Flasks
Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.
Manufacturer:  Corning™ 3275
 View more versions of this product
Catalog No. 07-200-75
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Medical-grade polystyrene is sterile and nonpyrogenic
  • Tissue culture-treated to promote optimal cell attachment and growth
  • Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
  • Graduated
  • Frosted labeling surfaces
  • Extra-wide neck openings for easier pipetting
  • Printed lot number and catalog number on each flask
  • Vented filter caps
  • Maximize gas exchange while minimizing the possibility of contamination
  • Hydrophobic 0.2µm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened
Specifications
Specifications
Black
75cm2
For optimal cell attachment and growth
Straight
Sterile
90mL
HDPE
Polystyrene
Yes
Triangular
Tissue culture treated