Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Costar™ Cell Culture Triangular Flasks

Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.

Specifications

Closure Material HDPE
Material Polystyrene
Surface Coating Tissue Culture Treated
For Use With (Application) For optimal cell attachment and growth
Shape Triangular
View More Specs

Products 2
Catalog Number Mfr. No. Closure Color Culture Area Neck Style Volume (Metric) Working Price Quantity & Availability  
Catalog Number Mfr. No. Closure Color Culture Area Neck Style Volume (Metric) Working Price Quantity & Availability  
07-200-66
View Documents
Corning™
3276
Orange Straight This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
07-200-63
View Documents
Corning™
3056
Orange Angled This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

 

  • Medical-grade polystyrene is sterile and nonpyrogenic
  • Tissue culture-treated to promote optimal cell attachment and growth
  • Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
  • Graduated
  • Frosted labeling surfaces
  • Extra-wide neck openings for easier pipetting
  • Printed lot number and catalog number on each flask
  • Vented filter caps
  • Maximize gas exchange while minimizing the possibility of contamination
  • Hydrophobic 0.2 µm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened

 

Specifications

Specifications

HDPE
Tissue Culture Treated
Triangular
Polystyrene
For optimal cell attachment and growth
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit