Learn More
Corning™ Costar™ Centrifugal Devices: Spin-X™ LC Available on GSA/VA Contract for Federal Government customers only.
Handles volumes up to 500µL. Corning™ Costar™ Centrifugal Devices: Spin-X™ LC includes a nylon membrane for applications such as HPLC sample preparation.
Manufacturer:  Corning™ 8169
 View more versions of this product
Catalog No. 07-200-389
Add to cart
Description
Description
  • Utilize centrifugation for the filtering of small volumes
  • Consist of a polypropylene filter insert and centrifuge tube
  • Handle volumes up to 500µL
  • Retained volume, <5µL
  • Withstand relative centrifugal force (RCF) to 16,000xG
Spin-X LC Units
  • Nylon membrane for applications such as HPLC sample preparation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Non-sterile
Nylon
0.22μm
Spin-X LC