Learn More
Corning™ Cluster Tube System Available on GSA/VA Contract for Federal Government customers only.
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Manufacturer:  Corning™ 4413
 View more versions of this product
Catalog No. 07-200-322
Add to cart
Description
Description
  • Convenient for large-volume transfer and collection from microtitration plates
  • Useful with single- and multichannel pipetters and in robotic workstations and centrifuges
  • 1.2mL individual tubes or breakaway strips of eight tubes; microtitration plate format
  • Assembled in 12- or 96-tube racks
  • Both tubes and racks are autoclavable clarified polypropylene
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sterile
Yes
Translucent
1mL
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges
Round
0.033 oz.
8-Strip Microplate Cluster Tube
Polypropylene
1.2mL
Rack
Strip
12 per rack
96
Documents
Accessories