Corning™ Costar™ Transwell™ Clear Polyester Membrane Inserts For 24-Well Plates Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Corning™ 3458
Catalog No. 09-762-25
  Check Availability
Add to Cart
    Specifications
    Surface Coating Cultrex Basement Membrane Extract
    Pore Density 1 x 10(5) pores/cm2
    Pore Size 0.4μm
    Type Membarne insert
    For Use With (Equipment) 24 Well plate
    Diameter (Metric) Filter 6.5mm
    Culture Area 0.33cm2
    Material PET
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback