Learn More
Corning™ Costar™ Transwell™-COL Collagen-Treated Coated Membrane Insert Available on GSA/VA Contract for Federal Government customers only.
Transparent, collagen-treated microporous membranes promote cell attachment and spreading
Manufacturer:  Corning™ 3492
 View more versions of this product
Catalog No. 07-200-559
Add to cart
Description
Description
  • Designed for transport studies, co-culture, and other experimental procedures; improves visibility of cells during culture
  • Membrane has an equimolar mixture of Types I and III collagen derived from bovine placenta—a biologically stabilized collagen which covers every fibril of the filter membrane matrix, maintaining porosity of the membrane
  • Inside wall of the insert has also been treated to allow formation of a complete and integral monolayer; can be viewed by phase-contrast microscopy
  • Tissue culture treated
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Individually packed and supplied in package with tissue culture plates with lids; sterile.
Specifications
Specifications
4.7cm2
Membrane Insert
24mm
3μm
6 Well Plate