Learn More
Corning™ Costar™ Transwell™-COL Collagen-Coated PTFE Membrane Insert
Transparent, collagen-treated microporous membranes promote cell attachment and spreading
Supplier: Corning™ 3493
Description
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
You can use cell culture inserts to:
- Produce a cell culture environment that closely resembles an in vivo state
- Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings Transparent, Collagen-coated PTFE membrane
- Promotes cell attachment and spreading
- Equimolar mixture of Collagen type I and III
- Individually packaged
- Multiple well plates included in each case
- Sterile
Order Info
Individually packed and supplied in package with tissue culture plates with lids; sterile.
Specifications
1.12 cm2 | |
1.5 mL | |
Membrane Insert | |
12 mm | |
24/Cs. | |
Sterile |
PTFE | |
0.4 μm | |
12 Well Plate | |
1/Pk. | |
Collagen | |
12 |