Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Costunolide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1mg
10mg
This item is not returnable. View return policy
553-21-9
C15H20O2
232.323
HRYLQFBHBWLLLL-BOHUOVHSSA-N
costunolide,unii-4ik578sa7z,costunlide,costunolid,costus lactone,ccris 6754,germacra-1 10 ,4,11 13-trien-12-oic acid, 6-alpha-hydroxy-, gamma-lactone, e,e,3as,10e,11ar-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca b furan-2-one,e,e-6-alpha-hydroxygermacra-1 10 ,4,11 13-trien-12-oic acid gamma-lactone,cyclodeca b furan-2 3h-one, 3a,4,5,8,9,11a-hexahydro-6,10-dimethyl-3-methylene-, 3as-3ar*,6e,10e,11as*
133640194
(3aS,6E,11aR)-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca[b]furan-2-one
CC1=CCCC(=CC2C(CC1)C(=C)C(=O)O2)C
This item is not returnable. View return policy
553-21-9
C15H20O2
232.323
HRYLQFBHBWLLLL-BOHUOVHSSA-N
costunolide,unii-4ik578sa7z,costunlide,costunolid,costus lactone,ccris 6754,germacra-1 10 ,4,11 13-trien-12-oic acid, 6-alpha-hydroxy-, gamma-lactone, e,e,3as,10e,11ar-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca b furan-2-one,e,e-6-alpha-hydroxygermacra-1 10 ,4,11 13-trien-12-oic acid gamma-lactone,cyclodeca b furan-2 3h-one, 3a,4,5,8,9,11a-hexahydro-6,10-dimethyl-3-methylene-, 3as-3ar*,6e,10e,11as*
133640194
(3aS,6E,11aR)-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca[b]furan-2-one
CC1=CCCC(=CC2C(CC1)C(=C)C(=O)O2)C

Inhibitor of human telomerase activity

Inhibitor of human telomerase activity (IC 50 = 65 μM in MCF-7 breast cancer cells). Suppresses proliferation and induces apoptosis in a variety of human tumor cell lines. Selectively blocks endothelial cell proliferation induced by VEGF. Inhibits expression of iNOS and IL-1β and disrupts NF-κB activation. Displays anti-inflammatory, antifungal and antiviral properties.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 1mg
Formula Weight 232.32
Percent Purity >97%
Chemical Name or Material Costunolide

Recommended Storage : Store at -20°C