Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Costunolide, Tocris Bioscience™

Click to view available options
Quantity:
1mg
10mg
This item is not returnable. View return policy
553-21-9
C15H20O2
232.323
HRYLQFBHBWLLLL-BOHUOVHSSA-N
costunolide,unii-4ik578sa7z,costunlide,costunolid,costus lactone,ccris 6754,germacra-1 10 ,4,11 13-trien-12-oic acid, 6-alpha-hydroxy-, gamma-lactone, e,e,3as,10e,11ar-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca b furan-2-one,e,e-6-alpha-hydroxygermacra-1 10 ,4,11 13-trien-12-oic acid gamma-lactone,cyclodeca b furan-2 3h-one, 3a,4,5,8,9,11a-hexahydro-6,10-dimethyl-3-methylene-, 3as-3ar*,6e,10e,11as*
133640194
(3aS,6E,11aR)-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca[b]furan-2-one
CC1=CCCC(=CC2C(CC1)C(=C)C(=O)O2)C
This item is not returnable. View return policy
553-21-9
C15H20O2
232.323
HRYLQFBHBWLLLL-BOHUOVHSSA-N
costunolide,unii-4ik578sa7z,costunlide,costunolid,costus lactone,ccris 6754,germacra-1 10 ,4,11 13-trien-12-oic acid, 6-alpha-hydroxy-, gamma-lactone, e,e,3as,10e,11ar-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca b furan-2-one,e,e-6-alpha-hydroxygermacra-1 10 ,4,11 13-trien-12-oic acid gamma-lactone,cyclodeca b furan-2 3h-one, 3a,4,5,8,9,11a-hexahydro-6,10-dimethyl-3-methylene-, 3as-3ar*,6e,10e,11as*
133640194
(3aS,6E,11aR)-6,10-dimethyl-3-methylidene-3a,4,5,8,9,11a-hexahydrocyclodeca[b]furan-2-one
CC1=CCCC(=CC2C(CC1)C(=C)C(=O)O2)C

Inhibitor of human telomerase activity

Inhibitor of human telomerase activity (IC 50 = 65 μM in MCF-7 breast cancer cells). Suppresses proliferation and induces apoptosis in a variety of human tumor cell lines. Selectively blocks endothelial cell proliferation induced by VEGF. Inhibits expression of iNOS and IL-1β and disrupts NF-κB activation. Displays anti-inflammatory, antifungal and antiviral properties.
Quantity 10mg
Formula Weight 232.32
Percent Purity >97%
Chemical Name or Material Costunolide

Recommended Storage : Store at -20°C