Learn More
BioVision IncSupplier Diversity Partner Cotinine ELISA Kit: ELISA kit that measure cotinine level, a reliable marker for smoking status and smoking cessation studies. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K4152100
Exposure to tobacco smoke can be detected by measuring nicotine and its metabolites. Nicotine has a short half life and is not used as a marker for tobacco smoke exposure. Cotinine due to its longer half life has been used in research as a reliable marker for smoking status and smoking cessation studies. BioVision’s Cotinine Direct ELISA Kit is designed for the detection Cotinine in serum and urine. It can also be adapted for other fluids. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-113-2370