Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Cotton Peptone 200, ultra-filtered (UF)

Gibco™ Cotton Peptone 200, Ultra-Filtered (UF) is processed to enhance carbohydrate concentrations, which is important for some mammalian protein glycosylation processes.

$206.00 - $1750.00

Specifications

For Use With (Application) Mammalian Cell Culture
Format Powder
Product Type Dehydrated culture media
Storage Requirements Store in cool, dry conditions in well-sealed containers, 2°C to 25°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
67-010-4
View Documents
Gibco™
670104
500 g
Each for $206.00
Add to cart
 
67-010-5
View Documents
Gibco™
670105
5 kg
Each for $1,750.00
Add to cart
 
Description

Description

Cotton 200, UF, is also part of the Peptone Preview Pak which contains:

  • Cotton 100, UF—rich in nucleoside content and digested to enhance peptide content. It is processed to provide higher concentrations of vitamins, which can help maintain performance in mammalian and microbial processes.
  • Wheat 100, UF—ultra-filtered supplement and good source of glutamine, which makes it ideal for mammalian cell culture applications. It is digested for enhanced peptide content resulting in a low concentration of free amino acids and carbohydrates.
  • Soy 100—highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates and is recommend for a wide variety of processes.
  • Pea 100, UF—ultra-filtered peptone digested to enhance nucleoside and free amino acid content. The high total amino acid content and low carbohydrate content are similar to many high-performance animal origin peptones.
  • Yeast 100—designed to enhance the content of vitamins and other key nutritional components found in yeast. It is intended for nutritional supplementation for mammalian, microbial, and insect cell culture.

Peptone Preview Pak supplements are non-GMP prototype products that are not for manufacture use. Larger sizes are available through the Gibco Custom Products Services.

Specifications

Specifications

Mammalian Cell Culture
Powder
Dehydrated culture media
Store in cool, dry conditions in well-sealed containers, 2°C to 25°C
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit