Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cottonseed Oil, NF, Spectrum™

Click to view available options
Quantity:
20 L
200 L
4 L
500 mL
Packaging:
Amber Glass Bottle
Drum
Poly Drum
8001-29-4
8001-29-4

CO145, 8001-29-4

Spectrum™ Cottonseed Oil, NF is used as a solvent and vehicle for injections, as an emollient vehicle for other medications and orally as a mild cathartic. It can also be used as a tablet binder for stable oral pharmaceutical powders. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 100%