Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cottonseed oil, pure, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
8001-29-4
MFCD00130872
8001-29-4
MFCD00130872
Density 0.9100g/mL
Color Yellow
Acidity 0.5mg KOH/g max.
Flash Point >112°C
Refractive Index 1.4720 to 1.4730
Specific Gravity 0.91
Quantity 500mL
Packaging Glass bottle
Saponification Value 185 to 198mg KOH/g
Infrared Spectrum Authentic
Merck Index 10,2535
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,benzene,chloroform and carbon,disulfide,slightly soluble in alcohol
Physical Form Oil
Grade Pure
Chemical Name or Material Cottonseed oil
Show More Show Less

LIGHT SENSITIVE

EINECSNumber : 232-280-7

RTECSNumber : GN2815000

TSCA : TSCA