Learn More
Coulter™ ImmunoPrep Reagent System
Rapid, no-wash, standardized whole blood lysing solution
$902.00 - $3740.00
Specifications
Type Immunoprep™ Reagent System
Format Liquid
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) No. of Tests Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) No. of Tests Price Quantity & Availability  
CO7546999
View Documents
Coulter™
7546999
300.0
Each for $3,740.00
Add to cart
CO7546946
View Documents
Coulter™
7546946
100.0
Each for $902.00
Add to cart
Description
Description
  • Superior sample to sample, laboratory to laboratory reproducibility
  • Gentle lysing action reduces cell stress and minimizes cell shrinkage while maintaining cellular integrity and morphology
  • Maximizes separation of cell populations of interest and red blood cells from debris
  • Optimizes scatter characteristics
Note: For In Vitro Diagnostic Use
Specifications
Specifications
Immunoprep™ Reagent System
Liquid