Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Coulter Whole Blood Lysing Reagent Kit
For the preparation of whole blood, bone marrow aspirates, and other sample types for analysis
$311.00 - $537.00
Specifications
Product Type | Whole Blood Lysing Reagents Kit |
---|---|
Includes | Immunolyse, fixative, and stabilizer solutions |
Certifications/Compliance | IVD, CE, FCM |
Includes: Immunolyse, Fixative, and Stabilizer solutions
Description
- 100test kit prepares samples for analysis by fluorescence microscopy or flow cytometry
- 300test kit prepares samples for flow cytometry only
For In Vitro Diagnostic Use
Specifications
Whole Blood Lysing Reagents Kit | |
Immunolyse, fixative, and stabilizer solutions | |
IVD, CE, FCM |
Safety and Handling
Recommended Storage : 2° to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title