Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COUP-TF I/NR2F1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | COUP-TF I/NR2F1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15282520
|
Novus Biologicals
NBP15282520UL |
20 μL |
Each for $204.00
|
|
|||||
NBP152825
|
Novus Biologicals
NBP152825 |
100 μL |
Each for $482.50
|
|
|||||
Description
COUP-TF I/NR2F1 Polyclonal specifically detects COUP-TF I/NR2F1 in Human samples. It is validated for Western Blot.Specifications
COUP-TF I/NR2F1 | |
Polyclonal | |
Rabbit | |
P10589 | |
7025 | |
Synthetic peptides corresponding to NR2F1(nuclear receptor subfamily 2, group F, member 1) The peptide sequence was selected from the N terminal of NR2F1. Peptide sequence GEQQQQAGSGAPHTPQTPGQPGAPATPGTAGDKGQGPPGSGQSQQHIECV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
COUP transcription factor 1, COUP-TF1, COUP-TFI, EAR-3COUP transcription factor I, EAR3COUP-TF I, ERBAL3V-erbA-related protein 3, Nuclear receptor subfamily 2 group F member 1, nuclear receptor subfamily 2, group F, member 1, SVP44, TCFCOUP1, TFCOUP1NR2F2, transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-ahomolog-like 3) | |
NR2F1 | |
IgG |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title