Learn More
Cole-Parmer Parker Hannifin Parker Straight Threaded Coupling, PFA, 1/4" NPT(F)  DFS Item
Manufacturer:  Cole-Parmer 3132060
Threaded fittings such as these offer more permanent installation.
Catalog No. 50-632-020