Learn More
Cole-Parmer CPC (Colder) SMCD01 Miniature Quick-Disconnect Coupling; Acetal; Valved, 1/16" ID  DFS Item
Manufacturer:  Cole-Parmer 0636387
Able to handle 1/16" ID tubing, these fittings offer a more secure connection alternative to luer fittings. Fitting halves engage and seal with a twist. Once sealed, the coupling allows tubing to rotate freely to help prevent kinked tubing and accidental disconnections.
Catalog No. 50-628-289