Learn More
Cole-Parmer CPC (Colder) MPCK32039 Sealing Cap with Lock, Polysulfone, 1/4", 5/pack  DFS Item
Manufacturer:  Cole-Parmer 3131177
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-908