Learn More
Cole-Parmer CPC (Colder) MCD1602 Metal Quick-Disconnect Panel-Mount Hose Barb Body; Valved, 1/8" Tube ID  DFS Item
Manufacturer:  Cole-Parmer 3140424
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180°F, and can be used with gases or liquids.
Catalog No. 50-627-840