Learn More
National Safety Apparel™ Coverall 7 oz. Navy Blue Indura Ultra Soft Zipper 32 in. Inseam
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability. With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort
$241.47 - $321.23
Specifications
Material Cotton, Nylon
Seam Type Stitched
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-469
View Documents
National Safety Apparel™
C88UJLZSM32
Small
Each for $241.47
Add to cart
17-001-470
View Documents
National Safety Apparel™
C88UJLZMD32
Medium
Each for $241.47
Add to cart
17-001-471
View Documents
National Safety Apparel™
C88UJLZLG32
Large
Each for $241.47
Add to cart
17-001-472
View Documents
National Safety Apparel™
C88UJLZXL32
X-Large
Each for $241.47
Add to cart
17-001-473
View Documents
National Safety Apparel™
C88UJLZ2X32
2X-Large
Each for $265.29
Add to cart
17-001-474
View Documents
National Safety Apparel™
C88UJLZ3X32
3X-Large
Each for $265.29
Add to cart
17-001-475
View Documents
National Safety Apparel™
C88UJLZ4X32
4X-Large
Each for $291.19
Add to cart
17-001-476
View Documents
National Safety Apparel™
C88UJLZ5X32
5X-Large
Each for $321.23
Add to cart
Description
Description
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 7 oz. Westex™ UltraSoft™
  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Home laundry
  • Made in the USA
Specifications
Specifications
Cotton, Nylon
Stitched