Learn More
National Safety Apparel™ Coverall 8 oz. Navy Blue Protera 30 in. Inseam
Coveralls with two-way covered zipper down front for easy entry
Manufacturer:  National Safety Apparel™ C88LY4X30
 View more versions of this product
Catalog No. 17-001-411
Add to cart
Description
Description
  • Constructed from Protera™
  • Comfortable, sewn-in elastic waist
  • Two-way covered zipper down front for easy entry
  • Breast pockets with flaps and hidden snap closures
  • Two large rear pockets
  • Two front pockets with pass-through slashes for access to pants pockets
  • All metal covered to ensure no con
Specifications
Specifications
12 cal CAT 2 ASTM F1506, NFPA 70 E, CSA Z462, OSHA 1910.269
Open
Protera 12 Cal Coverall
4X-Large
Zipper
Navy
Stitched
None