Learn More
National Safety Apparel™ Coverall 9 oz. Navy Blue Indura Ultra Soft Zipper 30 in. Inseam
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability. With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$261.13 - $348.17
Specifications
Material Cotton, Nylon
Seam Type Stitched
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-557
View Documents
National Safety Apparel™
C88UPLZSM30
Small
Each for $261.13
Add to cart
17-001-558
View Documents
National Safety Apparel™
C88UPLZMD30
Medium
Each for $261.13
Add to cart
17-001-559
View Documents
National Safety Apparel™
C88UPLZLG30
Large
Each for $261.13
Add to cart
17-001-560
View Documents
National Safety Apparel™
C88UPLZXL30
X-Large
Each for $261.13
Add to cart
17-001-561
View Documents
National Safety Apparel™
C88UPLZ2X30
2X-Large
Each for $288.08
Add to cart
17-001-562
View Documents
National Safety Apparel™
C88UPLZ3X30
3X-Large
Each for $288.08
Add to cart
17-001-563
View Documents
National Safety Apparel™
C88UPLZ4X30
4X-Large
Each for $316.06
Add to cart
17-001-564
View Documents
National Safety Apparel™
C88UPLZ5X30
5X-Large
Each for $348.17
Add to cart
Description
Description
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Zip front closure with additional covered snaps
  • Home laundry
  • Made in the USA
Specifications
Specifications
Cotton, Nylon
Stitched