Learn More
National Safety Apparel™ Coverall 9 oz. Navy Blue Indura Ultra Soft Zipper 32 in. Inseam
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability. With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$261.13 - $348.17
Specifications
Material Cotton, Nylon
Seam Type Stitched
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-565
View Documents
National Safety Apparel™
C88UPLZSM32
Small
Each for $261.13
Add to cart
17-001-566
View Documents
National Safety Apparel™
C88UPLZMD32
Medium
Each for $261.13
Add to cart
17-001-567
View Documents
National Safety Apparel™
C88UPLZLG32
Large
Each for $261.13
Add to cart
17-001-568
View Documents
National Safety Apparel™
C88UPLZXL32
X-Large
Each for $261.13
Add to cart
17-001-569
View Documents
National Safety Apparel™
C88UPLZ2X32
2X-Large
Each for $288.08
Add to cart
17-001-570
View Documents
National Safety Apparel™
C88UPLZ3X32
3X-Large
Each for $288.08
Add to cart
17-001-571
View Documents
National Safety Apparel™
C88UPLZ4X32
4X-Large
Each for $316.06
Add to cart
17-001-572
View Documents
National Safety Apparel™
C88UPLZ5X32
5X-Large
Each for $348.17
Add to cart
Description
Description
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Zip front closure with additional covered snaps
  • Home laundry
  • Made in the USA
Specifications
Specifications
Cotton, Nylon
Stitched