Learn More
Biotium CoverGrip™ Coverslip Sealant
Supplier: Biotium 23005
CoverGrip™ Coverslip Sealant is the first product specifically designed for sealing the edges of wet-mounted coverslips for fluorescence microscopy. Unlike nail polish, CoverGrip™ contains no ingredients that can leach into aqueous mounting medium and affect specimen fluorescence. CoverGrip™ is made with d-limonene, a natural, environmentally-friendly solvent with a pleasant citrus aroma, and dries to form a clear, hard, durable seal along the edges of the coverslip. Available in 15 mL brush-cap applicator bottles (23005) or in 100 mL dropper-cap refill bottles (23005-1).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.