Learn More
Covers and Gaskets for Nalgene™ Reusable Filter Funnel with Clamp Available on GSA/VA Contract for Federal Government customers only.
Accessories and replacement parts for reusable filter funnel.
Manufacturer:  Thermo Scientific 7103150004
 View more versions of this product
Catalog No. 08-645-31
Add to cart
Description
Description
Replace accessories and parts for the reusable filter funnel using these covers and gaskets. Continue to perform high throughput particle and microbiological QC testing using the Thermo Scientific™ Nalgene™ Reusable Filter Funnel with Clamp.
These are replacement parts and accessories for Nalgene™ Reusable Filter Funnel with Clamp, Cat #DS0315-0047
The Reusable Filter Funnel Features:
  • Tight fit prevents bypass even after autoclaving
  • Reservoir graduated from 100 to 250mL in 25mL increments
  • Aluminum clamp allows one-handed operation and easymembrane replacement
Includes: Rugged polysulfone funnel; polypropylene copolymer cover with three ports, three friction-fit TPE caps; support plate for 47mmmembrane; silicone gasket; No. 8 stopper (0.5625 in. hole); aluminum clamp
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Nalgene™ Reusable Filter Funnel
Accessory
Gasket