Learn More
Boson™ Covid-19 At Home Rapid Antigen Self Test
A highly accurate, rapid, at home self-use test for the detection of SARS-CoV-2 antigens in anterior nasal specimens. Includes 500 total tests (2 Tests/Pack, 250 Packs/Case).
Supplier: Boson™ 1N40C52US
Alert:
The Rapid SARS-CoV-2 Antigen Test Card is only for use under the Food and Drug Administration’s Emergency Use Authorization.
Description
- For Emergency Use Authorization use only
- Can be used on persons aged 2 years and older
- Results in 15 minutes
- Serial testing required
Specifications
Waived (FDA, EUA) | |
No | |
Self-collected anterior nasal (nares) swab samples | |
Covid-19 Self-Test | |
2 Rapid SARS-CoV-2 Antigen Test Cards; 2 Sterilied Swabs; 2 Extraction Buffer Tubes; Quick Reference Instructions and Pk. Insert; Built-in Tube Holder | |
Non-sterile |
SARS-CoV-2 | |
Cassette | |
Studies available upon request that show the tests sensitivity and specificity compared to other brands. | |
Single-use | |
2 Tests/Pk, 250 Pk/Cs, 500 Total Tests |