Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical Covidien™ Alcohol Prep Pads

Provides maximum absorbency for scrubbing and cleaning

Specifications

For Use With (Application) Provide maximum absorbency for scrubbing and cleaning
Sterility Sterile
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-898-098
View Documents
Moore Medical
34089
Medium This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description

Description

  • Non-woven sponge preps
  • Contains 70% isopropyl alcohol
  • Sterile
  • 2-ply

Specifications

Specifications

Provide maximum absorbency for scrubbing and cleaning
Sterile
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit