Learn More
Covidien Monoject™ 20cc Syringes
Bold graduations: 1cc
Manufacturer:  Covidien 8881520673
 View more versions of this product
Catalog No. 05-561-67
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Sterile latex-free syringes
  • Autoclavable in rigid pack for use in O.R/Procedure Packs
  • Robust compact package
Specifications
Specifications
Autoclavable
Yes
Monoject™ Standard Syringe
Sterile
For use in O.R./procedure packs
20 mL
1 mL
Regular luer-tip