Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ 6cc Syringes

Latex-free, bold graduations: 0.2cc

Specifications

Autoclavable Autoclavable
For Use With (Application) For use in O.R./procedure packs
Latex Free Yes
Product Type Monoject™ Standard Syringe
Graduations 0.2 mL
View More Specs

Products 2
Catalog Number Mfr. No. Connection Type Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type Price Quantity & Availability  
05-561-64
View Documents
Covidien
8881516911
Luer Tip
Case of 500 for $418.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
05-561-65
View Documents
Covidien
8881516937
Luer Lock
Case of 500 for $348.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Ultra-sharp, tribeveled, anticoring needle
  • Luer-Lok™ Syringe Tip on combination units
  • Color-coded translucent needle hub and rigid pack packing
  • Robust, compact package
  • Autoclavable in rigid pack for use in O.R./procedure packs

Specifications

Specifications

Autoclavable
Yes
0.2 mL
Tri-beveled, Anti-coring
For use in O.R./procedure packs
Monoject™ Standard Syringe
6 mL
Sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit