Learn More
Covidien Monoject™ Bone Marrow Biopsy Trays with Safety Components
Ideal for obtaining a reliable aspirate and/or core bone marrow sample with unaltered architecture
Manufacturer:  Covidien 8881847019
 View more versions of this product
Catalog No. 22-256622
Add to cart
Description
Description
  • Rigid stainless-steel needle features a double-cutting edge cannula and sharp style for confident use and smoother entry into the marrow
  • Comfortable ergonomic handle design provides precision control with minimal discomfort
  • Conveniently packaged in sterile procedural trays that comply with industry standards
Compliance
In compliance with the current FDA Quality System Requirements (QSR) as stipulated in 21 CFR Part 820.
Specifications
Specifications
16 (I-Type Sternal-Iliac); 11 (Bone Marrow)
Safety components, 16G I-type sternal-iliac needle and 11 gauge bone marrow needle
For bone marrow collection
Biopsy Tray