Learn More
Covidien Monoject™ Corvac™ Silicone-Coated Tubes with Red and Gray Mottled Stoppers
Integrated serum separator tubes
Manufacturer:  Covidien 8881302114
 View more versions of this product
Catalog No. 22-030579
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
12.5mL
Yes
0.63 in.
4.92 in.
16 x 125mm
Silicone coated; Gel barrier
Red and gray
16mm
125mm
Integrated serum separator tube