Learn More
Covidien Monoject™ Corvac™ Silicone-Coated Tubes with Red and Gray Mottled Stoppers
Integrated serum separator tubes
Manufacturer:  Covidien 8881302106
 View more versions of this product
Catalog No. 22-302106
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Improved performance and more sample yield for laboratory procedures requiring serum
  • Contains a positive gel barrier between blood cells and serum
  • Interior silicone glass powder activates the clotting of blood
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging; tubes are sterilized by gamma radiation
  • Crown Corvac™ gel placement for improved performance and more sample yield
  • TDM compatible
  • Allows for sensitive and routine pharmacokinetic testing
  • Plastic coating prevents tube cracking
  • Can be used with both angle-head and horizontal-head centrifuges
  • Laboratory procedures requiring serum with a positive gel barrier between blood cells and serum; general chemistry, serology and toxicology procedures
Specifications
Specifications
Sterile
Silicone coated; Gel barrier
0.51 in.
3.94 in.
13 x 100mm
6mL
Red and gray
13mm
100mm
Integrated serum separator tube