Learn More
Covidien Monoject™ Hypodermic Needles with Aluminum Hub
Color-coded, two-piece rigid pack sheath coupled with a precision aluminum hub
Manufacturer:  Covidien 8881200029
 View more versions of this product
Catalog No. 05-561-51
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Yes
Precision aluminum hub
1.5 in.
Stainless steel
Sterile
15 ga.
Provides very secure fit, needle can be manipulated with less chance of breakage compared to epoxy deisgn
Yes
38.1mm
Bevel
Hypodermic Needle