Learn More
Covidien Monoject™ MAGELLAN™ Safety Needles and Syringes
One-handed operation needle-based safety technology
Manufacturer:  Covidien 8881866215
 View more versions of this product
Catalog No. 22-256-559
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Yes
22 ga
6mL
Yes
3.81cm
Hypodermic Magellan™ Syringe
Tri-beveled, Anti-coring
Black
1.5 in.
6.0mL