Learn More
Covidien Monoject™ Nonsilicone-Coated Blood Collection Tubes with Green Stoppers
Blood-collection tubes with sterile interior
Manufacturer:  Covidien 8881320157
 View more versions of this product
Catalog No. 22-320157
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging
  • Sterilized by gamma radiation
  • Sodium heparin additive, 143 units
Laboratory procedures requiring plasma or whole blood; general hematology and certain chemistry procedures
Specifications
Specifications
Blood collection
sodium heparin 143 units
10.25 x 50mm
2mL draw
Green stopper
Glycerine stopper coating