Learn More
Covidien Monoject™ Nonsilicone Coated Tubes with Gray Stoppers
For laboratory procedures
Manufacturer:  Covidien 8881352481
 View more versions of this product
Catalog No. 22-352481
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging
  • Sterilized by gamma radiation
  • Used for laboratory procedures requiring plasma or whole blood and chemistry procedures where glycolytic inhibition of specimen is required
Specifications
Specifications
Blood collection
Powder additive: 10mg potassium oxalate, 12.5mg sodium fluoride
13 × 75mm (0.51 × 2.95 in.)
5mL draw
Gray stopper
Nonsilicone-Coated Tube with Gray Stopper