Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Rigid Pack 12mL Syringes

Convenient single-use individual packaging

Manufacturer:  Covidien 8881512860

 View more versions of this product

Catalog No. 19-038-499

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Specifications

Yes
Yes
0.2mL
Tri-beveled, Anti-coring
Eccentric, luer-tip
12mL
Luer tip
For use in O.R./procedure packs
Yes
Sterile
Monoject™ Syringe
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit