Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Safety Syringes: Insulin Syringes with Needles Attached

Design helps keep clinicians safe from accidental needlesticks before and after use

Manufacturer:  Covidien 8881511110

 View more versions of this product

Catalog No. 05-561-25



 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Specifications

OSHA
U-100 insulin
Yes
1.27cm
Sterile
29 ga.
Dark Orange
Yes
0.5 in.
Polypropylene
Monoject™ Insulin Safety Syringe
1mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit