Covidien Monoject™ SoftPack Hypodermic Needles

Color-coded gauge stripe and bar code on individual packages make it easy to identify needle sizes

Manufacturer: Covidien 1188822112

 View more versions of this product

Catalog No. 22-557-161

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Gauge 22 ga
    Length (English) Needle 1.5 in.
    Length (Metric) Needle 38.1mm
    Type Monoject™ Standard Hypodermic Needle
    For Use With (Application) Provides very secure fit, needle can be manipulated with less chance of breakage compared to epoxy deisgn
    Includes Polypropylene hub
    Latex Free Yes
    Material Stainless Steel
    Needle Point Style Regular Bevel
    Sterile Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit