Learn More
Covidien Monoject™ Supra/Hone™ Thin Wall Blood Collection Needles
Single- and multiple-sample needles from 20 to 22 gauge with color-coded sheaths
Manufacturer:  Covidien 8881216066
 View more versions of this product
Catalog No. 22-030573
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
Description
Description
  • Color-coded needle sheath designates the needle gauge
  • Unique polymer coating on stainless-steel needle enables easier puncture
  • One-piece molded valve on multiple sample needles prevent leakage
  • Single piece flow-through cannula design lessens the likelihood of hemolysis
Specifications
Specifications
22 G
Plastic hub
Monoject™ Blood Collection Needle
Sterile
Designates needle gauge, prevents leakage
1 in.
Stainless Steel