Covidien RCRA Hazardous Waste Containers

Effectively segregate hazardous pharmaceutical waste in accordance with the Resource Conservation and Recovery Act (RCRA)

Manufacturer: Covidien 8602RC

 View more versions of this product

Catalog No. 22-031-376

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance RCRA, EPA, DOT
    Closure Type Plain
    Color Black
    For Use With (Application) Segregate hazardous pharmaceutical waste under the Resource Conservation and Recovery Act
    Includes Snap cap
    Capacity (English) 2 gal.
    Product Type Waste Container

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit