Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Covidien RCRA Hazardous Waste Containers
Effectively segregate hazardous pharmaceutical waste in accordance with the Resource Conservation and Recovery Act (RCRA)
Supplier: Covidien 8608RC
Description
- Black container with white lid makes it easy to distinguish from sharps, chemo, and other disposal containers
- Leak-resistant gasket and absorbent pad help contain liquid contents
Compliance
RCRA Hazardous Waste Label meets EPA satellite accumulation labeling requirements. Meets DOT requirements for transport without secondary packaging.
Specifications
Waste Container | |
Slide Top | |
Black | |
Segregate hazardous pharmaceutical waste under the Resource Conservation and Recovery Act |
RCRA, EPA, DOT | |
8 gal. | |
Gasket, absorbent pad, and extra labels, sliding lid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction