Learn More
Covidien Sterile Specimen Containers
Meet CLSI standards
$216.30 - $265.51
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Includes | Price | Quantity & Availability | |||
14-375-152C
|
Covidien
2210SA |
2 oz. | 59.1mL | Metal lid and Plastisol™ lining |
Case of 200 for $216.30
|
|
14-375-152D
|
Covidien
2300SA |
4 oz. | 118mL | Metal lid and Plastisol™ lining, wide mouth, positive seal indicator |
Case of 200 for $265.51
|
|
Description
- Containers with metal lid and Plastisol lining minimize chances of leakage and prevent loss of specimen
- The clear plastic construction with graduations offers easy observation and measurement of contents
- The highly visible “positive seal” indicator clearly shows container is sealed when red bars are lined up
Specifications
CLSI | |
Plastic | |
Sterile |