Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education Preserved Specimens: Cow Organs
Larger specimens for easier dissection.
$58.00
Catalog Number | Product Type | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Product Type | Quantity | Price | Quantity & Availability | |||||
S1711S
![]() |
Beef Spinal Cord | Each |
Pack for $7.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S1715S
![]() |
Beef Brain | 15/CS |
Pack for $67.75
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S1722S
![]() |
Bull Testicle | Pack of 1 |
Pack for $15.75
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S8203S
![]() |
Beef Heart | Each |
Pack for $37.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S8202S
![]() |
Cow Eye | 10 Pack |
Pack of 10 for $32.75
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. |
|||||||||
S8207S
![]() |
Pregnant Cow uterus | 5/CS |
Each for $58.00
|
|
Description
- Organs are of excellent quality
- Large size makes them easy for students to dissect
- Vacuum packed in Fisher Science Education Bio-Fresh
Specifications
Bio-Fresh™ | |
Large |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title