Learn More
Abnova™ COX10 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001352T02
Catalog No. 89-940-260
Add to cart
Description
Description
Specifications
Specifications
Western Blot
COX10 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 48.84kDa
Transient overexpression cell lysate was tested with anti-COX10 antibody (Mfr. No. H00001352-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC000060.2
pCMV-COX10 full-length
100 μL
Human