Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX11 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA575268

Catalog No. PIPA575268


Only null left
Add to Cart

Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Cytochrome c oxidase (COX) is the terminal enzyme in the electron transfer chain, functioning as a transmembrane proton pump that builds an electrochemical gradient with chemical energy from the reduction of O2. Cytochrome c oxidase assembly protein COX11 is an intracellular mitochondrial membrane protein necessary for the construction of an active COX complex. COX11 contains a single transmembrane helix downstream of the N-terminal, mitochondrial targeting sequence and a C-terminal Cu(I)-binding domain. The assembly of COX requires the delivery of metal cofactors. Along with COX12 and SCO1/2, COX11 acts as a metal ion chaperone necessary for copper insertion into CuA and CuB redox-active copper centers of COX in eukaryotes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

COX11
Polyclonal
Unconjugated
COX11
2010004I09Rik; Cox11; COX11 cytochrome c oxidase copper chaperone; COX11 homolog, cytochrome c oxidase assembly protein; COX11, cytochrome c oxidase copper chaperone; COX11P; cytochrome c oxidase assembly homolog 11; cytochrome c oxidase assembly protein 11; cytochrome c oxidase assembly protein COX11, mitochondrial; cytochrome c oxidase subunit 11
Rabbit
Antigen affinity chromatography
RUO
1353, 690300, 69802
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Q6P8I6, Q9Y6N1
COX11
Synthetic peptide corresponding to amino acids 50-100 of Human COX11.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.