Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX15 Polyclonal Antibody
COX15 Polyclonal Antibody
Supplier: Thermo Scientific PA534681
Description
COX15 Polyclonal Antibody for Western Blot

Specifications
COX15 | |
Polyclonal | |
Unconjugated | |
COX15 | |
2900026G05Rik; CEMCOX2; COX15; COX15 cytochrome c oxidase assembly homolog; COX15 homolog, cytochrome c oxidase assembly protein; COX15, cytochrome c oxidase assembly homolog; cytochrome c oxidase assembly homolog 15; cytochrome c oxidase assembly homolog 15 (yeast); cytochrome c oxidase assembly protein 15; cytochrome c oxidase assembly protein COX15 homolog; cytochrome c oxidase subunit 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.52 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q7KZN9 | |
COX15 | |
Recombinant fragment corresponding to a region within amino acids 1 and 254 of COX15 (Uniprot ID#Q7KZN9). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction