Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX16 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550189

Catalog No. PA550189


Add to cart

Description

Description

The antibody detects endogenous levels of total COX16 protein.

This protein is encoded by COX16, corresponding to the previously uncharacterized open reading frame YJL003w of the yeast genome. COX16 was identified in studies of COX-deficient mutants previously assigned to complementation group G22 of a collection of yeast pet mutants. To determine its location, Cox16p was tagged with a Myc epitope at the C terminus.
Specifications

Specifications

COX16
Polyclonal
Unconjugated
COX16
Liquid
COX16
Fusion protein corresponding to a region derived from internal residues of human COX16 cytochrome c oxidase assembly homolog (S. cerevisiae)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9P0S2
C14orf112
Rabbit
IgG
100 μL
-20°C
Primary
51241
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit