Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA594950
Description
Synthetic peptide sequence: 26-41aa, CQNRGVCMSVGFDQYK. Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
COX2 (Cyclooxygenase 2, PTGS2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.
Specifications
COX2 | |
Polyclonal | |
0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl, 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal | |
P35354, P35355, Q05769 | |
Ptgs2 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human COX2, different from the related rat sequence by two amino acids. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PTGS2 | |
Prostaglandin G/H synthase 2; 1.14.99.1; Cyclooxygenase-2; COX-2; PHS II; Prostaglandin H2 synthase 2; PGH synthase 2; PGHS-2; Prostaglandin-endoperoxide synthase 2; PTGS2; COX2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19225, 29527, 5743 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction